Color-Coded Inserts
Supplier: SARSTEDT INC
65.744.999
77588-248CS 208.49 USD
77588-248
Color-Coded Inserts
Cap Inserts
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Suitable for: Screw caps of tubes 16.5 mm
  • Different colors for better coding
  • Quick and flexible coding and identification
  • Easy to push in
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.