Screw Cap with Retaining Loop
Supplier: SARSTEDT INC
65.712
77588-152CS 612.34 USD
77588-152
Screw Cap with Retaining Loop
Closures Test/Sample Tube Closures
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Polypropylene material
  • O-ring cap increases cap security (IATA tested)
  • Ribbed surface for maximum grip
  • Can be used on all SARSTEDT screw cap micro tubes
Attached cap ensures reliable assignment to the container. Color-coded inserts allow the sample to be identified quickly.