Screw Caps for Transport Containers, Round
Supplier: SARSTEDT INC
65.678 65.679 65.679.004 65.676 65.676.001
77588-142CS 85.44 USD
77588-142 77588-144 77588-146 77588-138 77588-140
Screw Caps for Transport Containers, Round
Closures Test/Sample Tube Closures
SARSTEDT offers sample transport systems and secondary containers for a wide variety of transport requirements.
  • Material: PP
  • Screw cap suitable for round mailing containers
  • Available in different colors
  • Autoclavable at 121 °C
Mailing containers with screw cap; a secure container for blood, urine and stool sample tubes.