Anti-Evaporation Cap for S-Monovette®
Supplier: SARSTEDT INC
65.648.100
77588-134CS 252.87 USD
77588-134
Anti-Evaporation Cap for S-Monovette®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation
The high quality of blood samples collected with the S-Monovette® blood collection system is due to the gentle aspiration technique. It has been proven to significantly reduce hemolysis rates compared to vacuum systems. As a result, erythrocytes remain intact and blood collection does not have to be repeated as often.
  • Suitable for S-Monovette® Ø 15 mm
  • Prevents unwanted evaporation of sample material
  • Prevents foreign particles from entering the sample material
  • Prevents contamination
Due to the gentle aspiration technique, the S-Monovette® blood collection system is suitable for all vein conditions, from young to old. It offers optimal sample quality and the best results thanks to the revolutionary blood collection system.
This also includes a well-designed, environmentally friendly packaging concept to reduce waste and a ready-to-use system for our needles. All needles can be used straight out of the packaging and allow users to work quickly - there is no assembly required as the needles are available exclusively with integrated holders.