In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, this range offers many products for collecting urine, stool, saliva, CSF and tissue samples.
- Design: With transport tube 30 ml
- Material: PE; Preparation: With stabilizer
- Volume: 3000 ml
- Heightר: 243×83.5 mm
- Complete set for reliable and patient-friendly urine collection over 24 hours
- 9 ml stabilizer (HCl, 20%)
- Design with light-protected 30 ml transport container
Easy-to-hold urine collection cup (75.1356) and self-adhesive label with information on patient and collection period enclosed. 3 L urine container, with light protection, with large opening diameter (80 mm), ergonomic recessed grips, transparent inspection strip and graduations.
Convenient and hygienic 24-hour urine collection.