Microvette® 500 Tubes with Collection Rim
Supplier: SARSTEDT INC
20.1341.102 20.1343.100 20.1344 20.1345.100 20.1346.100 20.1339.100
77587-442CS 708.23 USD
77587-442 77587-444 77587-446 77587-448 77587-450 77587-440
Microvette® 500 Tubes with Collection Rim
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Capillary blood collection facilitates diagnostics with a small volume of blood. This range offers innovative solutions for all patient groups.
  • Closure: Cap (HDPE)
  • Outer container with skirted base
  • Smooth cap – special lid design reduces the aerosol effect
  • Graduations to check fill level
  • Cylindrical inner tube: Optimal tube shape ensures the sample is well mixed
  • Collection via collection rim
The capillary blood collection systems are among the most versatile on the market. Microvette® with nominal volumes between 100 and 500 μl, conical or cylindrical inner shapes, as well as choice of collection technique - end-to-end or with a collection rim.