AccuTec Pro® Single Edge Blade, 3 Facet, Coated
Supplier: Accutec Blades
APBL-7063-0000
77587-138CS 2301.42 USD
77587-138
AccuTec Pro® Single Edge Blade, 3 Facet, Coated
Razors and Blades
AccuTec Pro® Single edge blades feature precision ground edges for sharpness and durability. The proprietary MicroCoat® Enrichment Technology reduces cut force and increases durability. Blades are wrapped in protective paper.
  • Blade edge with MicroCoat® technology enhances durability and sharpness
  • 3 facet edge for increased sharpness
  • High quality stainless steel blade
  • Blades are individually wrapped for safety
  • Reclosable clamshell for safe and convenient blade storage
  • Made in USA
The MicroCoat® blade enrichment technology adds a layer of protection for each blade, helping to maintain a sharp and effective edge over multiple uses.
Packaging: Each clamshell contains 100 blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.