Anti-NFKBIA Mouse Antibody (HRP (Horseradish Peroxidase))
Supplier: Sino Biological
12045MM02H100
77584-414EA 360.5 USD
77584-414
Anti-NFKBIA Mouse Antibody (HRP (Horseradish Peroxidase))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IkB alpha / NFKBIA (rh IkB alpha / NFKBIA; Catalog#12045-H07E; NP_065390.1; Phe 2-Leu 317). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).