Anti-LGALS9 Mouse Antibody
Supplier: Sino Biological
11147-MM11-100
77583-758EA 277.07 USD
77583-758
Anti-LGALS9 Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Galectin 9 (rh Galectin 9; Catalog#11147-H01H; NP_002299.2; Ala2-Thr323). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.