SteriPlast® Scraper, Detectable
Supplier: BURKLE INC
5378-2030
77581-236PK 49.84 USD
77581-236
SteriPlast® Scraper, Detectable
Scrapers
PS, blue. This scraper is not only colored blue but also has a special additive in the material that makes it visible to metal detectors or X rays. The product can therefore be rejected with standard systems for checking for foreign objects.
  • Disposable
  • Sturdy and convenient
  • Produced in a class seven cleanroom (10000)
  • Individually packaged and then sterilized by gamma rays
  • Can be detected by color scanners, metal detectors or X-rays
Suitable for scraping off, smoothing or cleaning as well as removing residue.