Cellulose Extraction Thimbles, Grade C300
Supplier: Sartorius
FT-1201-019090 FT-1201-022080 FT-1201-022100 FT-1201-025060 FT-1201-025070 FT-1201-025080 FT-1201-025100 FT-1201-026060 FT-1201-028060 FT-1201-028080 FT-1201-033205 FT-1201-035150 FT-1201-040100 FT-1201-040123 FT-1201-040150 FT-1201-043123 FT-1201-028100 FT-1201-030080 FT-1201-030100 FT-1201-033060 FT-1201-033080 FT-1201-033090 FT-1201-033094 FT-1201-033100 FT-1201-033118 FT-1201-033130
77581-142PK 112.75 USD
77581-142 77581-144 77581-146 77581-148 77581-150 77581-152 77581-154 77581-156 77581-158 77581-160 77581-182 77581-184 77581-186 77581-188 77581-190 77581-192 77581-162 77581-164 77581-166 77581-168 77581-170 77581-172 77581-174 77581-176 77581-178 77581-180
Cellulose Extraction Thimbles, Grade C300
Extractors Extraction Thimbles
These cellulose thimble are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives).
  • Composed of high-purity cellulose for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
They are free of binders and offer a high purity which ensures consistent and high overall flow rates.
Typical wall thickness are 1.3 mm for inner diameters lower than 35 mm and 1.7 mm for inner diameters higher than 35 mm.