InVivoSIM Anti-integrin Human Antibody

Supplier: Bio X Cell
SIM0025-100MG SIM0025-1MG SIM0025-25MG SIM0025-50MG SIM0025-5MG
77580-750EA 7984.56 USD
77580-750 77580-752 77580-754 77580-756 77580-758
InVivoSIM Anti-integrin Human Antibody
Antibodies
Guselkumab biosimilar.

Type:
Antigen: integrin
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR