InVivoSIM Anti-NGF Human Antibody

Supplier: Bio X Cell
SIM0017-100MG SIM0017-1MG SIM0017-25MG SIM0017-50MG SIM0017-5MG
77580-670EA 7984.56 USD
77580-670 77580-672 77580-674 77580-676 77580-678
InVivoSIM Anti-NGF Human Antibody
Antibodies
Tanezumab biosimilar.

Type:
Antigen: NGF
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR