InVivoSIM Anti-IL-17A Human Antibody

Supplier: Bio X Cell
SIM0013-100MG SIM0013-1MG SIM0013-25MG SIM0013-50MG SIM0013-5MG
77580-630EA 7984.56 USD
77580-630 77580-632 77580-634 77580-636 77580-638
InVivoSIM Anti-IL-17A Human Antibody
Antibodies
Secukinumab biosimilar.

Type:
Antigen: IL-17A
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR