RecombiMAb HIS Human IgG1 F(ab) Isotype Control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP179-100MG CP179-1MG CP179-25MG CP179-50MG CP179-5MG
77580-450EA 6269.61 USD
77580-450 77580-452 77580-454 77580-456 77580-458
RecombiMAb HIS Human IgG1 F(ab) Isotype Control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG1
Clonality: Recombinant
Clone: N/A-CP179
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR