RecombiMAb Human IgG1 (LALA-PG) Isotype control Anti-Respiratory Syncytial Virus

Supplier: Bio X Cell
CP161-100MG CP161-1MG CP161-25MG CP161-50MG CP161-5MG
77580-360EA 6269.61 USD
77580-360 77580-362 77580-364 77580-366 77580-368
RecombiMAb Human IgG1 (LALA-PG) Isotype control Anti-Respiratory Syncytial Virus
Antibodies
Type: Control
Antigen: IgG1
Clonality: Recombinant
Clone: Palivizumab-CP161
Conjugation:
Epitope:
Host:
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR