RecombiMAb Anti-PD-L1 Mouse Recombinant Antibody [clone: 10F.9G2™-CP158]

Supplier: Bio X Cell
CP158-100MG CP158-1MG CP158-25MG CP158-50MG CP158-5MG
77580-330EA 6269.61 USD
77580-330 77580-332 77580-334 77580-336 77580-338
RecombiMAb Anti-PD-L1 Mouse Recombinant Antibody [clone: 10F.9G2™-CP158]
Antibodies
Type: Primary
Antigen: PD-L1
Clonality: Recombinant
Clone: 10F.9G2™-CP158
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR