RecombiMAb Human IgG4 (S228P) Isotype control Anti-Respiratory Syncytial Virus

Supplier: Bio X Cell
CP152-100MG CP152-1MG CP152-25MG CP152-50MG CP152-5MG
77580-270EA 6269.61 USD
77580-270 77580-272 77580-274 77580-276 77580-278
RecombiMAb Human IgG4 (S228P) Isotype control Anti-Respiratory Syncytial Virus
Antibodies
Type: Control
Antigen: IgG4
Clonality: Recombinant
Clone: Palivizumab-CP152
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR