RecombiMAb Human IgG4 (S228P) Isotype Control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP147-100MG CP147-1MG CP147-25MG CP147-50MG CP147-5MG
77580-220EA 6269.61 USD
77580-220 77580-222 77580-224 77580-226 77580-228
RecombiMAb Human IgG4 (S228P) Isotype Control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG4
Clonality: Recombinant
Clone: N/A-CP147
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR