RecombiMAb Anti-CD71 Mouse Recombinant Antibody [clone: R17]

Supplier: Bio X Cell
CP130-100MG CP130-1MG CP130-25MG CP130-50MG CP130-5MG
77580-150EA 6269.61 USD
77580-150 77580-152 77580-154 77580-156 77580-158
RecombiMAb Anti-CD71 Mouse Recombinant Antibody [clone: R17]
Antibodies
Type: Primary
Antigen: CD71
Clonality: Recombinant
Clone: R17
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR