RecombiMAb Anti-CD28 Mouse Recombinant Antibody [clone: D665-CP043]

Supplier: Bio X Cell
CP043-1MG
77580-090EA 530.45 USD
77580-090
RecombiMAb Anti-CD28 Mouse Recombinant Antibody [clone: D665-CP043]
Antibodies
Type: Primary
Antigen: CD28
Clonality: Recombinant
Clone: D665-CP043
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR