RecombiMAb Anti-4-1BBL Mouse Recombinant Antibody [clone: TKS-1-CP041]

Supplier: Bio X Cell
CP041-1MG
77580-086EA 530.45 USD
77580-086
RecombiMAb Anti-4-1BBL Mouse Recombinant Antibody [clone: TKS-1-CP041]
Antibodies
Type: Primary
Antigen: 4-1BBL
Clonality: Recombinant
Clone: TKS-1-CP041
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR