RecombiMAb Anti-GITR Mouse Recombinant Antibody [clone: DTA-1-CP029]

Supplier: Bio X Cell
CP029-1MG
77580-070EA 530.45 USD
77580-070
RecombiMAb Anti-GITR Mouse Recombinant Antibody [clone: DTA-1-CP029]
Antibodies
Type: Primary
Antigen: GITR
Clonality: Recombinant
Clone: DTA-1-CP029
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR