RecombiMAb Anti-CTLA-4 Mouse Recombinant Antibody [clone: 9D9-CP006]

Supplier: Bio X Cell
CP006-1MG
77580-048EA 530.45 USD
77580-048
RecombiMAb Anti-CTLA-4 Mouse Recombinant Antibody [clone: 9D9-CP006]
Antibodies
Type: Primary
Antigen: CTLA-4
Clonality: Recombinant
Clone: 9D9-CP006
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR