InVivoPlus Mouse IgG2c Isotype control Anti-Dengue Virus

Supplier: Bio X Cell
BP0366-100MG BP0366-25MG BP0366-50MG BP0366-5MG
77580-032EA 5735.04 USD
77580-032 77580-034 77580-036 77580-038
InVivoPlus Mouse IgG2c Isotype control Anti-Dengue Virus
Antibodies
Type: Control
Antigen: IgG2c
Clonality:
Clone: DV5-1
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Dengue virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR