InVivoMAb Anti-CD205 Rat Monoclonal Antibody [clone: NLDC-145]

Supplier: Bio X Cell
BE0420-100MG BE0420-1MG BE0420-25MG BE0420-50MG BE0420-5MG
77579-740EA 4408.4 USD
77579-740 77579-742 77579-744 77579-746 77579-748
InVivoMAb Anti-CD205 Rat Monoclonal Antibody [clone: NLDC-145]
Antibodies
Type: Primary
Antigen: CD205
Clonality: Monoclonal
Clone: NLDC-145
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR