InVivoMAb Anti-CD28 Mouse Monoclonal Antibody [clone: JJ316]

Supplier: Bio X Cell
BE0418-100MG BE0418-1MG BE0418-25MG BE0418-50MG BE0418-5MG
77579-720EA 4408.4 USD
77579-720 77579-722 77579-724 77579-726 77579-728
InVivoMAb Anti-CD28 Mouse Monoclonal Antibody [clone: JJ316]
Antibodies
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: JJ316
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR