InVivoMAb Anti-CD2 Mouse Monoclonal Antibody [clone: OX-34]

Supplier: Bio X Cell
BE0417-100MG BE0417-1MG BE0417-25MG BE0417-50MG BE0417-5MG
77579-710EA 4408.4 USD
77579-710 77579-712 77579-714 77579-716 77579-718
InVivoMAb Anti-CD2 Mouse Monoclonal Antibody [clone: OX-34]
Antibodies
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: OX-34
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR