InVivoMAb Anti-CD8a Mouse Monoclonal Antibody [clone: OX-8]

Supplier: Bio X Cell
BE0415-100MG BE0415-1MG BE0415-25MG BE0415-50MG BE0415-5MG
77579-690EA 4408.4 USD
77579-690 77579-692 77579-694 77579-696 77579-698
InVivoMAb Anti-CD8a Mouse Monoclonal Antibody [clone: OX-8]
Antibodies
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: OX-8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR