InVivoMAb Anti-TCR α/β Mouse Monoclonal Antibody [clone: R73]

Supplier: Bio X Cell
BE0413-100MG BE0413-1MG BE0413-25MG BE0413-50MG BE0413-5MG
77579-670EA 4408.4 USD
77579-670 77579-672 77579-674 77579-676 77579-678
InVivoMAb Anti-TCR α/β Mouse Monoclonal Antibody [clone: R73]
Antibodies
Type: Primary
Antigen: TCR α/β
Clonality: Monoclonal
Clone: R73
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR