InVivoMAb Anti-GC1q R (C1QBP) Mouse Monoclonal Antibody [clone: 60.11]

Supplier: Bio X Cell
BE0411-100MG BE0411-1MG BE0411-25MG BE0411-50MG BE0411-5MG
77579-650EA 4408.4 USD
77579-650 77579-652 77579-654 77579-656 77579-658
InVivoMAb Anti-GC1q R (C1QBP) Mouse Monoclonal Antibody [clone: 60.11]
Antibodies
Type: Primary
Antigen: GC1q R (C1QBP)
Clonality: Monoclonal
Clone: 60.11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR