InVivoMAb Anti-EphA2 Mouse Monoclonal Antibody [clone: SHM16]

Supplier: Bio X Cell
BE0410-100MG BE0410-1MG BE0410-25MG BE0410-50MG BE0410-5MG
77579-640EA 4408.4 USD
77579-640 77579-642 77579-644 77579-646 77579-648
InVivoMAb Anti-EphA2 Mouse Monoclonal Antibody [clone: SHM16]
Antibodies
Type: Primary
Antigen: EphA2
Clonality: Monoclonal
Clone: SHM16
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR