InVivoMAb Anti-Fy3 Mouse Monoclonal Antibody [clone: MIMA-29]

Supplier: Bio X Cell
BE0407-100MG BE0407-1MG BE0407-25MG BE0407-50MG BE0407-5MG
77579-610EA 4408.4 USD
77579-610 77579-612 77579-614 77579-616 77579-618
InVivoMAb Anti-Fy3 Mouse Monoclonal Antibody [clone: MIMA-29]
Antibodies
Type: Primary
Antigen: Fy3
Clonality: Monoclonal
Clone: MIMA-29
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR