InVivoMAb Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 4H11-8]

Supplier: Bio X Cell
BE0401-100MG BE0401-1MG BE0401-25MG BE0401-50MG BE0401-5MG
77579-550EA 4408.4 USD
77579-550 77579-552 77579-554 77579-556 77579-558
InVivoMAb Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 4H11-8]
Antibodies
Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: 4H11-8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR