InVivoMAb Anti-CD83 Rat Monoclonal Antibody [clone: Michel-17]

Supplier: Bio X Cell
BE0398-100MG BE0398-1MG BE0398-25MG BE0398-50MG BE0398-5MG
77579-520EA 4408.4 USD
77579-520 77579-522 77579-524 77579-526 77579-528
InVivoMAb Anti-CD83 Rat Monoclonal Antibody [clone: Michel-17]
Antibodies
Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: Michel-17
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR