InVivoMAb Anti-MUC16 Mouse Monoclonal Antibody [clone: VK8]

Supplier: Bio X Cell
BE0391-100MG BE0391-1MG BE0391-25MG BE0391-50MG BE0391-5MG
77579-450EA 4408.4 USD
77579-450 77579-452 77579-454 77579-456 77579-458
InVivoMAb Anti-MUC16 Mouse Monoclonal Antibody [clone: VK8]
Antibodies
Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: VK8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR