InVivoMAb Anti-DKK3 Mouse Monoclonal Antibody [clone: DKK3-4.22]

Supplier: Bio X Cell
BE0385-100MG BE0385-1MG BE0385-25MG BE0385-50MG BE0385-5MG
77579-390EA 4408.4 USD
77579-390 77579-392 77579-394 77579-396 77579-398
InVivoMAb Anti-DKK3 Mouse Monoclonal Antibody [clone: DKK3-4.22]
Antibodies
Type: Primary
Antigen: DKK3
Clonality: Monoclonal
Clone: DKK3-4.22
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR