InVivoMAb Anti-GM-CSF Rat Monoclonal Antibody [clone: BVD2-23B6]

Supplier: Bio X Cell
BE0384-100MG BE0384-1MG BE0384-25MG BE0384-50MG BE0384-5MG
77579-380EA 4408.4 USD
77579-380 77579-382 77579-384 77579-386 77579-388
InVivoMAb Anti-GM-CSF Rat Monoclonal Antibody [clone: BVD2-23B6]
Antibodies
Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: BVD2-23B6
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR