InVivoMAb Anti-PD-L1 Mouse Monoclonal Antibody [clone: 368A.4H1]

Supplier: Bio X Cell
BE0383-100MG BE0383-1MG BE0383-25MG BE0383-50MG BE0383-5MG
77579-370EA 4408.4 USD
77579-370 77579-372 77579-374 77579-376 77579-378
InVivoMAb Anti-PD-L1 Mouse Monoclonal Antibody [clone: 368A.4H1]
Antibodies
Type: Primary
Antigen: PD-L1
Clonality: Monoclonal
Clone: 368A.4H1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse;Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR