InVivoMAb Anti-osteopontin (SPP1) Mouse Monoclonal Antibody [clone: MPIIIB10]

Supplier: Bio X Cell
BE0382-100MG BE0382-1MG BE0382-25MG BE0382-50MG BE0382-5MG
77579-360EA 4408.4 USD
77579-360 77579-362 77579-364 77579-366 77579-368
InVivoMAb Anti-osteopontin (SPP1) Mouse Monoclonal Antibody [clone: MPIIIB10]
Antibodies
Type: Primary
Antigen: osteopontin (SPP1)
Clonality: Monoclonal
Clone: MPIIIB10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse;Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR