InVivoMAb Anti-CD24 Rat Monoclonal Antibody [clone: M1/69]

Supplier: Bio X Cell
BE0360-100MG BE0360-1MG BE0360-25MG BE0360-50MG BE0360-5MG
77579-140EA 4408.4 USD
77579-140 77579-142 77579-144 77579-146 77579-148
InVivoMAb Anti-CD24 Rat Monoclonal Antibody [clone: M1/69]
Antibodies
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: M1/69
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR