InVivoMAb Anti-CA19-9 Mouse Monoclonal Antibody [clone: 1116-NS-19-9]

Supplier: Bio X Cell
BE0355-100MG BE0355-1MG BE0355-25MG BE0355-50MG BE0355-5MG
77579-090EA 4408.4 USD
77579-090 77579-092 77579-094 77579-096 77579-098
InVivoMAb Anti-CA19-9 Mouse Monoclonal Antibody [clone: 1116-NS-19-9]
Antibodies
Type: Primary
Antigen: CA19-9
Clonality: Monoclonal
Clone: 1116-NS-19-9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR