InVivoMAb Anti-ICOS Armenian hamster Monoclonal Antibody [clone: C398.4A]

Supplier: Bio X Cell
BE0353-100MG BE0353-1MG BE0353-25MG BE0353-50MG BE0353-5MG
77579-070EA 4408.4 USD
77579-070 77579-072 77579-074 77579-076 77579-078
InVivoMAb Anti-ICOS Armenian hamster Monoclonal Antibody [clone: C398.4A]
Antibodies
Type: Primary
Antigen: ICOS
Clonality: Monoclonal
Clone: C398.4A
Conjugation:
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: Mouse;Human;Rat;Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR