InVivoMAb Anti-PrP (Prion protein) Mouse Monoclonal Antibody [clone: TW1]

Supplier: Bio X Cell
BE0350-100MG BE0350-1MG BE0350-25MG BE0350-50MG BE0350-5MG
77579-040EA 4408.4 USD
77579-040 77579-042 77579-044 77579-046 77579-048
InVivoMAb Anti-PrP (Prion protein) Mouse Monoclonal Antibody [clone: TW1]
Antibodies
Type: Primary
Antigen: PrP (prion protein)
Clonality: Monoclonal
Clone: TW1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR