InVivoMAb Anti-TIM-4 Rat Monoclonal Antibody [clone: F31-5G3]

Supplier: Bio X Cell
BE0344-100MG BE0344-1MG BE0344-25MG BE0344-50MG BE0344-5MG
77578-990EA 4408.4 USD
77578-990 77578-992 77578-994 77578-996 77578-998
InVivoMAb Anti-TIM-4 Rat Monoclonal Antibody [clone: F31-5G3]
Antibodies
Type: Primary
Antigen: TIM-4
Clonality: Monoclonal
Clone: F31-5G3
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR