InVivoMAb Anti-CD71 Mouse Monoclonal Antibody [clone: OX-26]

Supplier: Bio X Cell
BE0331-100MG BE0331-1MG BE0331-25MG BE0331-50MG BE0331-5MG
77578-860EA 4408.4 USD
77578-860 77578-862 77578-864 77578-866 77578-868
InVivoMAb Anti-CD71 Mouse Monoclonal Antibody [clone: OX-26]
Antibodies
Type: Primary
Antigen: CD71
Clonality: Monoclonal
Clone: OX-26
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR