InVivoMAb Anti-CD28 Mouse Monoclonal Antibody [clone: D665]

Supplier: Bio X Cell
BE0328-100MG BE0328-1MG BE0328-25MG BE0328-50MG BE0328-5MG
77578-830EA 4408.4 USD
77578-830 77578-832 77578-834 77578-836 77578-838
InVivoMAb Anti-CD28 Mouse Monoclonal Antibody [clone: D665]
Antibodies
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: D665
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR