InVivoMAb Anti-IL-15 Rat Monoclonal Antibody [clone: AIO.3]

Supplier: Bio X Cell
BE0315-10MG
77578-808EA 1068.11 USD
77578-808
InVivoMAb Anti-IL-15 Rat Monoclonal Antibody [clone: AIO.3]
Antibodies
Type: Primary
Antigen: IL-15
Clonality: Monoclonal
Clone: AIO.3
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR