InVivoMAb Anti-CD16/CD32 Rat Monoclonal Antibody [clone: 2.4G2]

Supplier: Bio X Cell
BE0307-1MG
77578-804EA 169.85 USD
77578-804
InVivoMAb Anti-CD16/CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Antibodies
Type: Primary
Antigen: CD16/CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR