NANOCOLOR® Photometric Tube Tests, Sludge Activity
Supplier: CTL SCIENTIFIC SUPPLY CORP
985890
77578-596EA 350.2 USD
77578-596
NANOCOLOR® Photometric Tube Tests, Sludge Activity
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of sludge activity in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.